Learn More
Medchemexpress LLC Gallic acid (3,4,5-Trihydroxybenzoic acid) | 149-91-7 | 170.12 | 1 G
SDP
Catalog No. 5000389019
Encompass_Preferred
Gallic acid (3,4,5-Trihydroxybenzoic acid) is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). It possesses various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties.
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Antimicrobial activity
  • Antioxidant activity
  • Anti-inflammatory activity
  • Anticancer activity
Catalog No. 50-003-89019 Supplier Medchemexpress LLC Supplier No. HYN05231G
Gallic acid (3,4,5-Trihydroxybenzoic acid) is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). It possesses various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties.
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Antimicrobial activity
  • Antioxidant activity
  • Anti-inflammatory activity
  • Anticancer activity