Learn More
Medchemexpress LLC Gallic acid (3,4,5-Trihydroxybenzoic acid) | 149-91-7 | 170.12 | 5 G
SDP
Catalog No. 5000390122
Encompass_Preferred
Gallic acid is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). It possesses various activities including antimicrobial, antioxidant, anti-inflammatory, and anticancer activities.
  • Natural polyhydroxyphenolic compound.
  • Free radical scavenger.
  • Inhibits cyclooxygenase-2 (COX-2).
  • Demonstrates antimicrobial activity.
  • Possesses antioxidant properties.
  • Exhibits anti-inflammatory effects.
  • Shows anticancer activities.
  • Improves glucose tolerance and triglyceride concentration in diet-induced obesity mice.
  • Dramatically decreases the number of viable neutrophils after treatment.
Catalog No. 50-003-90122 Supplier Medchemexpress LLC Supplier No. HYN05235G
Gallic acid is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). It possesses various activities including antimicrobial, antioxidant, anti-inflammatory, and anticancer activities.
  • Natural polyhydroxyphenolic compound.
  • Free radical scavenger.
  • Inhibits cyclooxygenase-2 (COX-2).
  • Demonstrates antimicrobial activity.
  • Possesses antioxidant properties.
  • Exhibits anti-inflammatory effects.
  • Shows anticancer activities.
  • Improves glucose tolerance and triglyceride concentration in diet-induced obesity mice.
  • Dramatically decreases the number of viable neutrophils after treatment.