Learn More
Medchemexpress LLC Gallic acid-d2 (3,4,5-Trihydroxybenzoic acid-d2) | 294660-92-7 | 172.13 | 5 MG
SDP
Catalog No. 5000389021
Encompass_Preferred
Gallic acid-d2 is the deuterium labeled Gallic acid. Gallic acid (3,4,5-Trihydroxybenzoic acid) is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). Gallic acid exhibits various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer activities.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89021 Supplier Medchemexpress LLC Supplier No. HYN0523S5MG
Add to Cart
Add to Cart
Gallic acid-d2 is the deuterium labeled Gallic acid. Gallic acid (3,4,5-Trihydroxybenzoic acid) is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). Gallic acid exhibits various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer activities.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.