Learn More
Medchemexpress LLC Gallic acid-d2 50mg
SDP
Catalog No. 5000669526
Encompass_Preferred
Gallic acid-d2 is the deuterium labeled Gallic acid[1] Gallic acid (3 4 5-Trihydroxybenzoic acid) is a natural polyhydroxyphenolic compound and an free radical scavenger to inhibit cyclooxygenase-2 (COX-2)[2] Gallic acid has various activities such as antimicrobial antioxidant antimicrobial anti-inflammatory and anticance activities[3]
Catalog No. 50-006-69526 Supplier Medchemexpress LLC Supplier No. 50MGHYN0523S
Gallic acid-d2 is the deuterium labeled Gallic acid[1] Gallic acid (3 4 5-Trihydroxybenzoic acid) is a natural polyhydroxyphenolic compound and an free radical scavenger to inhibit cyclooxygenase-2 (COX-2)[2] Gallic acid has various activities such as antimicrobial antioxidant antimicrobial anti-inflammatory and anticance activities[3]