Learn More
Medchemexpress LLC HY-N0523A 100mg Medchemexpress, Gallic acid (hydrate) CAS:5995-86-8 Purity:>98%
SDP
Catalog No. 502082799
Encompass_Preferred
Medchemexpress, HY-N0523A 100mg Gallic acid (hydrate) CAS:5995-86-8 Gallic acid (3,4,5-Trihydroxybenzoic acid) hydrate is a natural polyhydroxyphenolic compound and an free radical scavenger to inhibit cyclooxygenase-2 (COX-2). Gallic acid hydrate has various activities, such as antimicrobial, antioxidant, antimicrobial, anti-inflammatory, and anticance activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2799 Supplier Medchemexpress LLC Supplier No. HYN0523A100MG
Medchemexpress, HY-N0523A 100mg Gallic acid (hydrate) CAS:5995-86-8 Gallic acid (3,4,5-Trihydroxybenzoic acid) hydrate is a natural polyhydroxyphenolic compound and an free radical scavenger to inhibit cyclooxygenase-2 (COX-2). Gallic acid hydrate has various activities, such as antimicrobial, antioxidant, antimicrobial, anti-inflammatory, and anticance activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.