Learn More
Medchemexpress LLC Gallic acid hydrate 1g
SDP
Catalog No. 5000669539
Encompass_Preferred
Gallic acid (3 4 5-Trihydroxybenzoic acid) hydrate is a natural polyhydroxyphenolic compound and an free radical scavenger to inhibit cyclooxygenase-2 (COX-2)[1] Gallic acid hydrate has various activities such as antimicrobial antioxidant antimicrobial anti-inflammatory and anticance activities[2]
Catalog No. 50-006-69539 Supplier Medchemexpress LLC Supplier No. 1GHYN0523A
Gallic acid (3 4 5-Trihydroxybenzoic acid) hydrate is a natural polyhydroxyphenolic compound and an free radical scavenger to inhibit cyclooxygenase-2 (COX-2)[1] Gallic acid hydrate has various activities such as antimicrobial antioxidant antimicrobial anti-inflammatory and anticance activities[2]