Learn More
Medchemexpress LLC Gallic acid hydrate | 5995-86-8 | 188.13 | 1 ML
SDP
Catalog No. 5000389751
Encompass_Preferred
Gallic acid hydrate (3,4,5-Trihydroxybenzoic acid hydrate) is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). It possesses various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties. It is intended for research use only and not for human use.
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial activity
  • Shows antioxidant activity
  • Possesses anti-inflammatory activity
  • Demonstrates anticancer activity
  • Improves glucose tolerance and triglyceride concentration in diet-induced obesity mice
Catalog No. 50-003-89751 Supplier Medchemexpress LLC Supplier No. HYN0523A10MM/1ML
Gallic acid hydrate (3,4,5-Trihydroxybenzoic acid hydrate) is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). It possesses various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties. It is intended for research use only and not for human use.
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial activity
  • Shows antioxidant activity
  • Possesses anti-inflammatory activity
  • Demonstrates anticancer activity
  • Improves glucose tolerance and triglyceride concentration in diet-induced obesity mice