Learn More
Hach Company Gallic Acid Solution, 50 mL SCDB
Supplier:  Hach Company 1442326
Encompass
Gallic acid Solution for conductivity testing. Used to neutralize hydroxide ion interference.
Catalog No. 50-199-5028