Learn More
Medchemexpress LLC Gallic acid (Standard) | 149-91-7 | 170.12 | 100 MG
SDP
Catalog No. 5000389355
Encompass_Preferred
Gallic acid (Standard) is the analytical standard of Gallic acid, intended for research and analytical applications. It is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2), exhibiting various activities such as antimicrobial, antioxidant, anti-inflammatory, and anticancer properties.
  • Analytical standard for gallic acid
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial, antioxidant, anti-inflammatory, and anticancer activities
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-89355 Supplier Medchemexpress LLC Supplier No. HYN0523R100MG
Gallic acid (Standard) is the analytical standard of Gallic acid, intended for research and analytical applications. It is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2), exhibiting various activities such as antimicrobial, antioxidant, anti-inflammatory, and anticancer properties.
  • Analytical standard for gallic acid
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial, antioxidant, anti-inflammatory, and anticancer activities
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS