Learn More
Medchemexpress LLC Gallic acid (Standard) | 149-91-7 | 170.12 | 25 MG
SDP
Catalog No. 5000389561
Encompass_Preferred
Gallic acid (Standard) is the analytical standard of Gallic acid, intended for research and analytical applications. It is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). This compound is an analytical standard commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Gallic acid exhibits various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties.
  • Analytical standard for research and analytical applications
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial activity
  • Exhibits antioxidant activity
  • Exhibits anti-inflammatory activity
  • Exhibits anticancer activity
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-003-89561 Supplier Medchemexpress LLC Supplier No. 25MGHYN0523R
Gallic acid (Standard) is the analytical standard of Gallic acid, intended for research and analytical applications. It is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2). This compound is an analytical standard commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Gallic acid exhibits various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties.
  • Analytical standard for research and analytical applications
  • Natural polyhydroxyphenolic compound
  • Free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial activity
  • Exhibits antioxidant activity
  • Exhibits anti-inflammatory activity
  • Exhibits anticancer activity
  • Used in HPLC, GC, and MS research experiments