Learn More
Medchemexpress LLC Gallic acid (Standard) | 149-91-7 | 170.12 | 50 MG
SDP
Catalog No. 5000389752
Encompass_Preferred
Gallic acid (Standard) is the analytical standard of Gallic acid, intended for research and analytical applications. It is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2).
  • Gallic acid has various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer activities.
  • The compound is an analytical standard, used as a reference standard assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-89752 Supplier Medchemexpress LLC Supplier No. HYN0523R50MG
Gallic acid (Standard) is the analytical standard of Gallic acid, intended for research and analytical applications. It is a natural polyhydroxyphenolic compound and a free radical scavenger that inhibits cyclooxygenase-2 (COX-2).
  • Gallic acid has various activities, including antimicrobial, antioxidant, anti-inflammatory, and anticancer activities.
  • The compound is an analytical standard, used as a reference standard assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.