Learn More
Selleck Chemical LLC Gallic acid trimethyl ether S3862-25mg
SDP
Catalog No. 503109672
Encompass_Preferred
Gallic acid trimethyl ether (Eudesmic acid Tri-O-methylgallic acid Benzoic acid 3 4 5-Trimethoxybenzoic acid) is an O-methylated trihydroxybenzoic acid that is found in olive
Catalog No. 50-310-9672 Supplier Selleck Chemical LLC Supplier No. S386225MG
Gallic acid trimethyl ether (Eudesmic acid Tri-O-methylgallic acid Benzoic acid 3 4 5-Trimethoxybenzoic acid) is an O-methylated trihydroxybenzoic acid that is found in olive