Learn More
Medchemexpress LLC Gallic aldehyde | 13677-79-7 | 154.12 | 1 G
SDP
Catalog No. 5000447643
Encompass_Preferred
Gallic aldehyde (3,4,5-Trihydroxybenzaldehyde) is a phenolic aldehyde isolated from Geum japonicum. It inhibits the gelatinolytic activity and expression of MMP-9, as well as ERK1/2, p38, and JNK. It also demonstrates potent anti-HSV-1 and antioxidant activities, and antibacterial activity against Oenococcus oeni VF.
  • Inhibits gelatinolytic activity and expression of MMP-9.
  • Inhibits ERK1/2, p38, and JNK.
  • Possesses potent anti-HSV-1 activity.
  • Exhibits antioxidant activities.
  • Demonstrates antibacterial activity against Oenococcus oeni VF.
Catalog No. 50-004-47643 Supplier Medchemexpress LLC Supplier No. HYW0044861G
Gallic aldehyde (3,4,5-Trihydroxybenzaldehyde) is a phenolic aldehyde isolated from Geum japonicum. It inhibits the gelatinolytic activity and expression of MMP-9, as well as ERK1/2, p38, and JNK. It also demonstrates potent anti-HSV-1 and antioxidant activities, and antibacterial activity against Oenococcus oeni VF.
  • Inhibits gelatinolytic activity and expression of MMP-9.
  • Inhibits ERK1/2, p38, and JNK.
  • Possesses potent anti-HSV-1 activity.
  • Exhibits antioxidant activities.
  • Demonstrates antibacterial activity against Oenococcus oeni VF.