Learn More
Medchemexpress LLC Gallic aldehyde | 13677-79-7 | 154.12 | 100 MG
SDP
Catalog No. 5000448803
Encompass_Preferred
Gallic aldehyde is a phenolic aldehyde that can be isolated from *Geum japonicum*. It has potent anti-HSV-1 and antioxidant activities. It also exhibits antibacterial activity against *Oenococcus oeni VF*.
  • Inhibits the gelatinolytic activity and expression of MMP-9.
  • Inhibits ERK1/2, P38, and JNK.
  • Potent anti-HSV-1 activity.
  • Antioxidant activity.
  • Antibacterial activity against *Oenococcus oeni VF*.
  • For research use only.
Catalog No. 50-004-48803 Supplier Medchemexpress LLC Supplier No. HYW004486100MG
Gallic aldehyde is a phenolic aldehyde that can be isolated from *Geum japonicum*. It has potent anti-HSV-1 and antioxidant activities. It also exhibits antibacterial activity against *Oenococcus oeni VF*.
  • Inhibits the gelatinolytic activity and expression of MMP-9.
  • Inhibits ERK1/2, P38, and JNK.
  • Potent anti-HSV-1 activity.
  • Antioxidant activity.
  • Antibacterial activity against *Oenococcus oeni VF*.
  • For research use only.