Learn More
Medchemexpress LLC Gallic aldehyde | 13677-79-7 | 154.12 | 250 MG
SDP
Catalog No. 5000449956
Encompass_Preferred
Gallic aldehyde is a phenolic aldehyde isolated from *Geum japonicum*. It is known for its inhibitory activity against gelatinolytic activity and expression of MMP-9. It also inhibits ERK1/2, p38, and JNK. This compound demonstrates potent anti-HSV-1 and antioxidant activities, alongside antibacterial activity against *Oenococcus oeni* VF.
  • Inhibits gelatinolytic activity and expression of MMP-9
  • Inhibits ERK1/2, p38, and JNK
  • Exhibits potent anti-HSV-1 activity
  • Demonstrates antioxidant activities
  • Shows antibacterial activity against *Oenococcus oeni* VF
  • For solid, store at 4°C under nitrogen
  • For solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-004-49956 Supplier Medchemexpress LLC Supplier No. 250MGHYW004486
Gallic aldehyde is a phenolic aldehyde isolated from *Geum japonicum*. It is known for its inhibitory activity against gelatinolytic activity and expression of MMP-9. It also inhibits ERK1/2, p38, and JNK. This compound demonstrates potent anti-HSV-1 and antioxidant activities, alongside antibacterial activity against *Oenococcus oeni* VF.
  • Inhibits gelatinolytic activity and expression of MMP-9
  • Inhibits ERK1/2, p38, and JNK
  • Exhibits potent anti-HSV-1 activity
  • Demonstrates antioxidant activities
  • Shows antibacterial activity against *Oenococcus oeni* VF
  • For solid, store at 4°C under nitrogen
  • For solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen