Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium ICP Standard, 10,000 ppm Ga in 5% HNO3, Ricca Chemical
SDP

Catalog No. PGA10KN50 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PGA10KN-50 50 mL Natural Poly Bottle
PGA10KN-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PGA10KN-50 Supplier Ricca Chemical Company Supplier No. PGA10KN50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-55-3 , 7697-37-2 , 7732-18-5
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Packaging Natural Poly Bottle
CAS Min % 75.0, 22.47, 0.75
CAS Max % 78.0, 23.39, 0.78
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 50 mL
Concentration or Composition (by Analyte or Components) 1mL = 10mg Ga (10, 000ppm Ga)Ga in 5% HNO3
Chemical Name or Material Gallium ICP Standard
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.