Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(II) chloride, ultra dry, 99.999% (metals basis)

Catalog No. AA3569603
Change view
Click to view available options
Quantity:
1 g
Packaging:
Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA3569603 1 g Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3569603 Supplier Thermo Scientific Chemicals Supplier No. 03569603
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24597-12-4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24597-12-4
MDL Number MFCD00151526

Specifications

Boiling Point 535°C
Melting Point 164°C
Color White
Physical Form Powder
Quantity 1 g
UN Number UN3260
Formula Weight 281.26
Percent Purity 99.999%
Odor Acidic
Packaging Ampule
Sensitivity Air and moisture sensitive
Assay (metals basis)
Chemical Name or Material Gallium(II) chloride, Ultra dry
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 246-338-4
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.