Learn More
Gallium(III) chloride, ultra dry, 99.999% (metals basis)
Description
Gallium(III) chloride is a catalyst for a number of transformations including the ring opening and recyclization of epoxides and the trimerization of alkynes. It is used to detect the solar neutrions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 201°C |
| Melting Point | 78°C |
| Physical Form | Granules |
| Quantity | 25 g |
| Packaging | Ampouled under argon |
| Sensitivity | Moisture sensitive |
| Specific Gravity | 2.47 |
| Density | 2.47 |
| UN Number | UN3260 |
| Solubility Information | Soluble in water,benzene,carbon tetrachloride and carbon disulfide |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 236-610-0 |
| RTECSNumber | LW9100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.