Learn More
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics™
Description
Gallium(III) nitrate hydrate, 99.9998%, (trace metal basis), ACROS Organics, Quantity: 100g, Packaging: Glass bottle, CAS: 69365-72-6, White, Grade: Trace Metal Basis, Melting Point: 110.0 deg.C, Molecular Formula: GaN3O9.xH2O, Molecular Weight: 255.74, Percent Purity: 99.9998%
for ultra trace analysisSpecifications
Assay Percent Range | 99.9998% |
Linear Formula | Ga(NO3)3·xH2O |
Merck Index | 14, 4350 |
Solubility Information | Solubility in water: soluble |
Formula Weight | 255.74 |
Physical Form | Crystalline Powder, Crystals, and/or Chunks |
Percent Purity | 99.9998% |
Grade | Trace Metal Basis |
Packaging | Glass bottle |
Solubility | Solubility in water: soluble. |
Show More |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger