Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gallium(III) oxide

Catalog No. 21506610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-506-610G 10 g
21-506-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-506-610G Supplier Sigma Aldrich Supplier No. 21506610G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Starting material for the preparation of Sr2CuGaO3S, an example of a rare square pyramidal gallium.

Specifications

CAS 12024-21-4
Quantity 10 g
Molecular Formula Ga2O3
Linear Formula Ga2O3
MDL Number MFCD00011020
Synonym Gallium trioxide
Molecular Weight (g/mol) 187.44
Percent Purity ≥99.99% trace metals basis
Density 5.88 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.