Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium ingot, 99.99% (metals basis)

Catalog No. AA1018818
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1018818 50 g
AA1018830 250 g
AA1018836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1018818 Supplier Thermo Scientific Chemicals Supplier No. 01018818
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-55-3 | Ga | 69.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Gray-Silver
Quantity 50 g
Form Ingot
UN Number UN2803
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Gallium ingot
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H290
May be corrosive to metals.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: GALLIUM
EINECSNumber 231-163-8
RTECSNumber LW8600000
TSCA Yes
storageNote1 Requires overnight shipping with cold pack; Store at 2°C to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.