Learn More
Medchemexpress LLC HY-N0521 1mg Medchemexpress, (-)-Gallocatechin CAS:3371-27-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05211MG
Encompass_Preferred
Medchemexpress, HY-N0521 1mg (-)-Gallocatechin CAS:3371-27-5 (-)-Gallocatechin, an epimer of (-)-Epigallocatechin (EGC), is contained in various tea products. (-)-Gallocatechin has antioxidant activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2798