Learn More
TARGETMOL CHEMICALS INC Gallocatechin 25MG
SDP
Catalog No. 502879171
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. (+)-Gallocatechin (Gallocatechol) possesses free radical scavenging ability. (+)-Gallocatechin has moderate affinity to the human cannabinoid receptor. Purity 98.12%
Catalog No. 50-287-9171 Supplier TARGETMOL CHEMICALS INC Supplier No. T384525MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. (+)-Gallocatechin (Gallocatechol) possesses free radical scavenging ability. (+)-Gallocatechin has moderate affinity to the human cannabinoid receptor. Purity 98.12%