Learn More
Medchemexpress LLC (-)-Gallocatechin | 3371-27-5 | 306.27 | 1 ML
SDP
Catalog No. 5000390225
Encompass_Preferred
(-)-Gallocatechin is an epimer of (-)-Epigallocatechin (EGC) found in various tea products. It demonstrates significant antioxidant activity in vitro through several mechanisms.
  • Epimer of (-)-Epigallocatechin (EGC).
  • Demonstrates antioxidant activity in vitro.
  • Inhibits LDL oxidation.
  • Scavenges DPPH radicals.
  • Inhibits thiobarbituric acid-reactive substances (TBARS) production.
  • Exhibits antioxidant activity in the Ferric reducing-antioxidant power (FRAP) assay.
Catalog No. 50-003-90225 Supplier Medchemexpress LLC Supplier No. HYN052110MM/1ML
(-)-Gallocatechin is an epimer of (-)-Epigallocatechin (EGC) found in various tea products. It demonstrates significant antioxidant activity in vitro through several mechanisms.
  • Epimer of (-)-Epigallocatechin (EGC).
  • Demonstrates antioxidant activity in vitro.
  • Inhibits LDL oxidation.
  • Scavenges DPPH radicals.
  • Inhibits thiobarbituric acid-reactive substances (TBARS) production.
  • Exhibits antioxidant activity in the Ferric reducing-antioxidant power (FRAP) assay.