Learn More
Medchemexpress LLC (-)-Gallocatechin | 3371-27-5 | 306.27 | 5 MG
SDP
Catalog No. 5000389237
Encompass_Preferred
(-)-Gallocatechin is an epimer of (-)-Epigallocatechin (EGC) found in various tea products. It demonstrates antioxidant activity in vitro by inhibiting LDL oxidation and scavenging DPPH radicals.
  • Inhibits production of thiobarbituric acid-reactive substances (TBARS) in Cu+2 mediated oxidation of human LDL.
  • Exhibits antioxidant activity in the Ferric reducing-antioxidant power (FRAP) assay.
  • Shows DPPH radical scavenging activity.
Catalog No. 50-003-89237 Supplier Medchemexpress LLC Supplier No. HYN05215MG
(-)-Gallocatechin is an epimer of (-)-Epigallocatechin (EGC) found in various tea products. It demonstrates antioxidant activity in vitro by inhibiting LDL oxidation and scavenging DPPH radicals.
  • Inhibits production of thiobarbituric acid-reactive substances (TBARS) in Cu+2 mediated oxidation of human LDL.
  • Exhibits antioxidant activity in the Ferric reducing-antioxidant power (FRAP) assay.
  • Shows DPPH radical scavenging activity.