Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-gallocatechin | 3371-27-5 | MFCD01632616 | 100.0% | 306.27 g/mol | C15H14O7 | 10 MG
SDP

Catalog No. 5000298045
Encompass_Preferred

(-)-Gallocatechin is a tea-derived flavanol (polyphenol) with antioxidant activity, provided as a high-purity analytical reagent for laboratory research. It is an epimer of (-)-epigallocatechin and is commonly used in biochemical and analytical studies.

  • Antioxidant polyphenol found in green tea.
  • Epimer of (-)-epigallocatechin (EGC).
  • CAS number 3371-27-5.
  • Chemical formula C15H14O7; molecular weight 306.27 g/mol.
  • Purity reported as 99.98%.
  • Storage: 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Pack quantity: 10 MG.

Catalog No. 50-002-98045 Supplier Medchemexpress LLC Supplier No. HYN052110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-Gallocatechin is a tea-derived flavanol (polyphenol) with antioxidant activity, provided as a high-purity analytical reagent for laboratory research. It is an epimer of (-)-epigallocatechin and is commonly used in biochemical and analytical studies.

  • Antioxidant polyphenol found in green tea.
  • Epimer of (-)-epigallocatechin (EGC).
  • CAS number 3371-27-5.
  • Chemical formula C15H14O7; molecular weight 306.27 g/mol.
  • Purity reported as 99.98%.
  • Storage: 4°C, protect from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Pack quantity: 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.