Learn More
Medchemexpress LLC --Gallocatechin 50mg
SDP
Catalog No. 5000669453
Encompass_Preferred
(-)-Gallocatechin an epimer of (-)-Epigallocatechin (EGC) (HY-N0225) is contained in various tea products (-)-Gallocatechin demonstrates antioxidant activity in vitro including inhibition of LDL oxidation and DPPH radical scavenging[1][2][3]
Catalog No. 50-006-69453 Supplier Medchemexpress LLC Supplier No. 50MGHYN0521
(-)-Gallocatechin an epimer of (-)-Epigallocatechin (EGC) (HY-N0225) is contained in various tea products (-)-Gallocatechin demonstrates antioxidant activity in vitro including inhibition of LDL oxidation and DPPH radical scavenging[1][2][3]