Learn More
Medchemexpress LLC (-)-Gallocatechin gallate | 4233-96-9 | 458.37 | 1 ML
SDP
Catalog No. 5000389238
Encompass_Preferred
(-)-Gallocatechin gallate is a polyphenol isolated from tea, with known cancer-preventive activities. This product is for research use only.
  • Inhibits G6PD-mediated NADPH production in mouse 3T3-L1 cells.
  • Shows synergistic effects on inducing apoptosis and inhibiting cell growth in PC-9 cells.
  • Exhibits inhibitory effects on alpha-Glucosidase and DPPH.
Catalog No. 50-003-89238 Supplier Medchemexpress LLC Supplier No. HYN052210MM/1ML
(-)-Gallocatechin gallate is a polyphenol isolated from tea, with known cancer-preventive activities. This product is for research use only.
  • Inhibits G6PD-mediated NADPH production in mouse 3T3-L1 cells.
  • Shows synergistic effects on inducing apoptosis and inhibiting cell growth in PC-9 cells.
  • Exhibits inhibitory effects on alpha-Glucosidase and DPPH.