Learn More
Medchemexpress LLC (-)-Gallocatechin gallate | 4233-96-9 | MFCD00214298 | 100.0% | 458.37 g/mol | C22H18O11 | 10 MG
SDP
Catalog No. 5000299015
Encompass_Preferred
(-)-Gallocatechin gallate is a tea-derived polyphenol supplied as an analytical standard for research. It exhibits antioxidant and reported cancer-preventive activity, and has been associated with effects on catechol-O-methyltransferase (COMT). Supplied as a high-purity white to off-white solid for analytical and biological research.
  • Tea-derived polyphenol with antioxidant properties.
  • Reported cancer-preventive activity and COMT-related effects.
  • High purity analytical standard (99.97% LCMS).
  • White to off-white solid, suitable for analytical use.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
Catalog No. 50-002-99015 Supplier Medchemexpress LLC Supplier No. HYN052210MG
(-)-Gallocatechin gallate is a tea-derived polyphenol supplied as an analytical standard for research. It exhibits antioxidant and reported cancer-preventive activity, and has been associated with effects on catechol-O-methyltransferase (COMT). Supplied as a high-purity white to off-white solid for analytical and biological research.
  • Tea-derived polyphenol with antioxidant properties.
  • Reported cancer-preventive activity and COMT-related effects.
  • High purity analytical standard (99.97% LCMS).
  • White to off-white solid, suitable for analytical use.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).