Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gallocatechin gallate | 5127-64-0 | 1 MG
SDP

Catalog No. 5000417843
Encompass_Preferred

Gallocatechin gallate is a flavonoids product that can be isolated from the herbs of Celastrus orbiculatus. It is intended for research use only.

  • Molecular weight: 458.37
  • Formula: C22H18O11
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98%
  • Storage: -20°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light).
  • Structure classification: Flavonoids
  • Initial source: Plants, celastraceae, celastrus, Celastrus orbiculatus Thunb.

Catalog No. 50-004-17843 Supplier Medchemexpress LLC Supplier No. HYN77271MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gallocatechin gallate is a flavonoids product that can be isolated from the herbs of Celastrus orbiculatus. It is intended for research use only.

  • Molecular weight: 458.37
  • Formula: C22H18O11
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98%
  • Storage: -20°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light).
  • Structure classification: Flavonoids
  • Initial source: Plants, celastraceae, celastrus, Celastrus orbiculatus Thunb.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.