Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gallocatechin gallate | 5127-64-0 | 10 MG
SDP

Catalog No. 5000417681
Encompass_Preferred

Gallocatechin gallate is a Flavonoids product isolated from the herbs of *Celastrus orbiculatus*. This compound has a molecular weight of 458.37 and a chemical formula of C22H18O11. It appears as a solid, white to off-white in color, and is suitable for research applications.

  • Isolated from *Celastrus orbiculatus*
  • Purity of 98%
  • Molecular weight of 458.37
  • Chemical formula C22H18O11
  • Solid appearance
  • White to off-white color
  • Classified as a flavonoid
  • For research use only

Catalog No. 50-004-17681 Supplier Medchemexpress LLC Supplier No. HYN772710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gallocatechin gallate is a Flavonoids product isolated from the herbs of *Celastrus orbiculatus*. This compound has a molecular weight of 458.37 and a chemical formula of C22H18O11. It appears as a solid, white to off-white in color, and is suitable for research applications.

  • Isolated from *Celastrus orbiculatus*
  • Purity of 98%
  • Molecular weight of 458.37
  • Chemical formula C22H18O11
  • Solid appearance
  • White to off-white color
  • Classified as a flavonoid
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.