Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gallocatechin gallate | 5127-64-0 | 5 MG
SDP

Catalog No. 5000419083
Encompass_Preferred

Gallocatechin gallate is a flavonoid product isolated from the herbs of Celastrus orbiculatus. This chemical compound is provided as a solid, typically white to off-white in appearance, with a purity of 98%.

  • Molecular weight: 458.37
  • Chemical formula: C22H18O11
  • Recommended storage at -20°C, sealed, away from moisture and light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • For research use only

Catalog No. 50-004-19083 Supplier Medchemexpress LLC Supplier No. HYN77275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gallocatechin gallate is a flavonoid product isolated from the herbs of Celastrus orbiculatus. This chemical compound is provided as a solid, typically white to off-white in appearance, with a purity of 98%.

  • Molecular weight: 458.37
  • Chemical formula: C22H18O11
  • Recommended storage at -20°C, sealed, away from moisture and light
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.