Learn More
AdipoGen (-)-Gallocatechin gallate (5 mg)
SDP
Catalog No. 502054463
Encompass_Preferred
Chemical. CAS: 4233-96-9. Formula: C22H18O11. MW: 458.37. (-)-Gallocatechin gallate is a catechin polyphenol that has been found in green tea extracts. It has been shown to have diverse biological activities, including antioxidant, antiallergic, antiobesity and anticancer properties. In addition it has been shown to have antibacterial and antitrypanosomal effects.
Catalog No. 50-205-4463 Supplier AdipoGen Supplier No. CDXG0197M005
Chemical. CAS: 4233-96-9. Formula: C22H18O11. MW: 458.37. (-)-Gallocatechin gallate is a catechin polyphenol that has been found in green tea extracts. It has been shown to have diverse biological activities, including antioxidant, antiallergic, antiobesity and anticancer properties. In addition it has been shown to have antibacterial and antitrypanosomal effects.