Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Gallocatechin gallate, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014178 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4178 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4178 Supplier MilliporeSigma Supelco Supplier No. 8035210MG
Add to Cart
Add to Cart
Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 4233-96-9
Physical Form Neat
Linear Formula C22H18O11
MDL Number MFCD00214298
Synonym (2 S,3 R)-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2 H)-benzopyran-3,5,7-triol 3-(3,4,5-trihydroxybenzoate)
Type Analytical Standard
Quantity 10 mg
Formula Weight 458.37
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.