Learn More
Selleck Chemical LLC Gallocyanine
SDP
Catalog No. 5000003573
Encompass_Preferred
Gallocyanine (Solid Violet) is a histological dye used in solution with an iron alum mordant as a hematoxylin substitute in the H E stain It has a role as a fluorochrome and a histological dye
Catalog No. 50-000-03573 Supplier Selleck Chemical LLC Supplier No. S560225MG
Gallocyanine (Solid Violet) is a histological dye used in solution with an iron alum mordant as a hematoxylin substitute in the H E stain It has a role as a fluorochrome and a histological dye