Learn More
Medchemexpress LLC Gallocyanine (chloride) | 1562-85-2 | 92.0% | 25 MG
SDP
Catalog No. 5000413561
Encompass_Preferred
Gallocyanine chloride is a synthetic blue dyestuff being investigated as a potential agent for Alzheimer's disease and related neurodegenerative tauopathies. It functions by inhibiting DKK1/LRP6 interaction with an IC50 of 6.38 μM, which in turn activates the Wnt signaling pathway and leads to β-catenin accumulation. This compound also demonstrates anti-metastasis, anti-inflammatory, and anti-fibrosis activities. Additionally, Gallocyanine chloride can be employed as a fluorescent probe for detecting superoxide anion radicals.
  • Potential research agent for Alzheimer's disease and neurodegenerative tauopathies
  • Inhibits DKK1/LRP6 interaction
  • Activates Wnt signaling pathway
  • Causes β-catenin accumulation
  • Exhibits anti-metastasis activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-fibrosis activity
  • Can be used as a fluorescent probe for superoxide anion radical detection
Catalog No. 50-004-13561 Supplier Medchemexpress LLC Supplier No. 25MGHYD0961
Gallocyanine chloride is a synthetic blue dyestuff being investigated as a potential agent for Alzheimer's disease and related neurodegenerative tauopathies. It functions by inhibiting DKK1/LRP6 interaction with an IC50 of 6.38 μM, which in turn activates the Wnt signaling pathway and leads to β-catenin accumulation. This compound also demonstrates anti-metastasis, anti-inflammatory, and anti-fibrosis activities. Additionally, Gallocyanine chloride can be employed as a fluorescent probe for detecting superoxide anion radicals.
  • Potential research agent for Alzheimer's disease and neurodegenerative tauopathies
  • Inhibits DKK1/LRP6 interaction
  • Activates Wnt signaling pathway
  • Causes β-catenin accumulation
  • Exhibits anti-metastasis activity
  • Exhibits anti-inflammatory activity
  • Exhibits anti-fibrosis activity
  • Can be used as a fluorescent probe for superoxide anion radical detection