Learn More
Medchemexpress LLC Gallocyanine chloride | 1562-85-2 | 92.0% | 5 MG
SDP
Catalog No. 5000410870
Encompass_Preferred
Gallocyanine chloride is a synthetic blue dyestuff used in research for Alzheimer's disease and related neurodegenerative tauopathies. It inhibits DKK1/LRP6 interaction and activates the Wnt signaling pathway.
  • Activates the Wnt signaling pathway
  • Causes β-catenin accumulation
  • Exhibits anti-metastasis, anti-inflammatory, and anti-fibrosis activities
  • Can serve as a fluorescent probe for detecting superoxide anion radicals
Catalog No. 50-004-10870 Supplier Medchemexpress LLC Supplier No. HYD09615MG
Gallocyanine chloride is a synthetic blue dyestuff used in research for Alzheimer's disease and related neurodegenerative tauopathies. It inhibits DKK1/LRP6 interaction and activates the Wnt signaling pathway.
  • Activates the Wnt signaling pathway
  • Causes β-catenin accumulation
  • Exhibits anti-metastasis, anti-inflammatory, and anti-fibrosis activities
  • Can serve as a fluorescent probe for detecting superoxide anion radicals