Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallocyanine 90.0+%, TCI America™
SDP

Catalog No. G017825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G017825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G017825G Supplier TCI America Supplier No. G017825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1562-85-2
Molecular Formula C15H13ClN2O5
Molecular Weight (g/mol) 336.728
MDL Number MFCD00011926
InChI Key AQSOTOUQTVJNMY-UHFFFAOYSA-N
Synonym gallocyanine, gallocyanin, solid violet, 1-carboxy-7-dimethylamino-3,4-dihydroxyphenoxazin-5-ium chloride, gallocyanine bs, gallocyanine dh, fast violet, anthracene blue swgg, alizarine navy blue at, brilliant chrome blue p
PubChem CID 73801
IUPAC Name 7-(dimethylamino)-4-hydroxy-3-oxophenoxazin-10-ium-1-carboxylic acid;chloride
SMILES CN(C)C1=CC2=C(C=C1)[NH+]=C3C(=CC(=O)C(=C3O2)O)C(=O)O.[Cl-]

Specifications

Quantity 25 g
Formula Weight 336.73
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gallocyanine
RTECSNumber SP7692000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.