Learn More
Selleck Chemical LLC Galloflavin S9606-5MG
SDP
Catalog No. 5000024253
Encompass_Preferred
Galloflavin (NSC 107022) is a novel inhibitor of lactate dehydrogenasee (LDH) with Ki values of 5 46 M and 15 06 M for LDH-A and LDH-B respectively
Catalog No. 50-000-24253 Supplier Selleck Chemical LLC Supplier No. S96065MG
Galloflavin (NSC 107022) is a novel inhibitor of lactate dehydrogenasee (LDH) with Ki values of 5 46 M and 15 06 M for LDH-A and LDH-B respectively