Learn More
US Plastic Corporation 1 Gallon Natural HDPE Round Jug with 38/400 Neck
SDP
Catalog No. NC1805578
Encompass
These are heavier weight, high-density polyethylene (HDPE), narrow-mouth containers. They have built-in handles for convenient usage and a funnel top for easy pouring. They are ideal for storing or transporting a variety of liquids and they meet FDA standards. *Does not include caps. Caps are sold separately
Catalog No. NC1805578 Supplier US Plastic Corporation Supplier No. 66244
These are heavier weight, high-density polyethylene (HDPE), narrow-mouth containers. They have built-in handles for convenient usage and a funnel top for easy pouring. They are ideal for storing or transporting a variety of liquids and they meet FDA standards. *Does not include caps. Caps are sold separately