Learn More
Aviva Systems Biology Corp GALM Antibody - middle region (ARP52649_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52649_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GALM. Alternate names: BLOCK 25, IBD1, BLOCK25ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-747