Learn More
Medchemexpress LLC GalNac-siRNA negative control (21nt) | 5 MG
SDP
Catalog No. 5000375004
Encompass_Preferred
GalNac-siRNA negative control (21nt) is the negative control form of GalNac-siRNA. GalNac-siRNA is an Asialoglycoprotein receptor (ASGPR)-targeted inhibitor conjugate, promising for research of hereditary transthyretin amyloidosis, acute hepatic porphyria, hemophilia, and hypercholesterolemia.
  • Oligonucleotides
  • SiRNAs
  • SiRNA drugs
Catalog No. 50-003-75004 Supplier Medchemexpress LLC Supplier No. HY150151A5MG
GalNac-siRNA negative control (21nt) is the negative control form of GalNac-siRNA. GalNac-siRNA is an Asialoglycoprotein receptor (ASGPR)-targeted inhibitor conjugate, promising for research of hereditary transthyretin amyloidosis, acute hepatic porphyria, hemophilia, and hypercholesterolemia.
  • Oligonucleotides
  • SiRNAs
  • SiRNA drugs