Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GalNac-siRNA negative control | 97.0% | 10 MG
SDP

Catalog No. 5000286230
Encompass_Preferred

GalNac-siRNA negative control (21nt) is a GalNAc-conjugated, 21-nucleotide non-targeting siRNA supplied as a solid white to off-white powder for use as a negative control in liver-directed RNA interference research. The oligonucleotide includes a GalNAc ligand for asialoglycoprotein receptor (ASGPR)-mediated hepatocyte delivery. Sequence: SS: UUCUCCGAACGUGUCACGUTT; AS: ACGUGACACGUUCGGAGAATT.

  • 21-nucleotide non-targeting siRNA.
  • GalNAc conjugation enables ASGPR-mediated liver delivery.
  • Intended for use as a negative control in RNAi experiments.
  • Purity approximately 97.0%.
  • Physical state: solid, white to off-white.
  • Storage: -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small-scale pack sizes such as 1 mg, 5 mg, 10 mg, and 50 mg.

Catalog No. 50-002-86230 Supplier Medchemexpress LLC Supplier No. HY150151A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GalNac-siRNA negative control (21nt) is a GalNAc-conjugated, 21-nucleotide non-targeting siRNA supplied as a solid white to off-white powder for use as a negative control in liver-directed RNA interference research. The oligonucleotide includes a GalNAc ligand for asialoglycoprotein receptor (ASGPR)-mediated hepatocyte delivery. Sequence: SS: UUCUCCGAACGUGUCACGUTT; AS: ACGUGACACGUUCGGAGAATT.

  • 21-nucleotide non-targeting siRNA.
  • GalNAc conjugation enables ASGPR-mediated liver delivery.
  • Intended for use as a negative control in RNAi experiments.
  • Purity approximately 97.0%.
  • Physical state: solid, white to off-white.
  • Storage: -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small-scale pack sizes such as 1 mg, 5 mg, 10 mg, and 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.