Learn More
Cayman Chemical GalNacL96trIethylamIn sal 10mg
SDP
Catalog No. 502945779
Encompass_Preferred
A triantennary GalNAc ASGPR ligand; has been used in the synthesis of siRNA-GalNAc conjugates for RNA interference-mediated gene silencing in vitro and in mice
Catalog No. 50-294-5779 Supplier Cayman Chemical Supplier No. 4045610 MG
A triantennary GalNAc ASGPR ligand; has been used in the synthesis of siRNA-GalNAc conjugates for RNA interference-mediated gene silencing in vitro and in mice