Learn More
Cayman Chemical GalNac-L96 (triethylamine salt), Purity: ≥95%, CAS: 1159408-62-4, 50 mg
SDP
Catalog No. 502945652
Encompass_Preferred
A triantennary GalNAc ASGPR ligand; has been used in the synthesis of siRNA-GalNAc conjugates for RNA interference-mediated gene silencing in vitro and in mice
Catalog No. 50-294-5652 Supplier Cayman Chemical Supplier No. 4045650 MG
A triantennary GalNAc ASGPR ligand; has been used in the synthesis of siRNA-GalNAc conjugates for RNA interference-mediated gene silencing in vitro and in mice