Learn More
Cayman Chemical Galanin (2-11) amide (human, mouse, rat, porcine, bovine, ovine) (trifluoroacetate salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502950172
Encompass_Preferred
A synthetic peptide fragment of galanin and an agonist of the GAL2 receptor (EC50 = 9.32 nM in a fluorescence imaging plate reader assay); selectively binds to the GAL2 receptor (IC50 = 1.76 nM for the rat receptor) over the GAL1 receptor (IC50 = 879 nM for the human receptor) but does also bind to the GAL3 receptor (Ki = 271 nM for the rat receptor); decreases immobility in the forced swim test in rats at 1 nmol/animal, i.c.v.; reduces the hind paw mechanical pain threshold and increases the hind paw cold sensitivity threshold in rats
Catalog No. 50-295-0172 Supplier Cayman Chemical Supplier No. 340651 MG
A synthetic peptide fragment of galanin and an agonist of the GAL2 receptor (EC50 = 9.32 nM in a fluorescence imaging plate reader assay); selectively binds to the GAL2 receptor (IC50 = 1.76 nM for the rat receptor) over the GAL1 receptor (IC50 = 879 nM for the human receptor) but does also bind to the GAL3 receptor (Ki = 271 nM for the rat receptor); decreases immobility in the forced swim test in rats at 1 nmol/animal, i.c.v.; reduces the hind paw mechanical pain threshold and increases the hind paw cold sensitivity threshold in rats