Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Galnon TFA | 1217448-19-5 | MFCD06798339 | 98.9% | 792.84 | C42H47F3N4O8 | 5 MG
SDP

Catalog No. 5000248197
Encompass_Preferred

Galnon TFA is a selective, non-peptide agonist of galanin receptors GAL1 and GAL2 used in neuroscience research. It binds GAL1 and GAL2 with reported Kis of 11.7 μM and 34.1 μM and has demonstrated anticonvulsant and anxiolytic effects. The compound is supplied as a white to off-white solid with high purity and is soluble in DMSO for in vitro use.

  • Selective galanin receptor agonist (GAL1 and GAL2).
  • Demonstrated anticonvulsant and anxiolytic activity in preclinical models.
  • High purity (98.9%) and white to off-white solid appearance.
  • Soluble in DMSO (125 mg/mL); use fresh DMSO and ultrasonic assistance if needed.
  • Recommended storage sealed, protected from moisture and light; frozen solution stability varies by temperature.

Catalog No. 50-002-48197 Supplier Medchemexpress LLC Supplier No. HY103536A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Galnon TFA is a selective, non-peptide agonist of galanin receptors GAL1 and GAL2 used in neuroscience research. It binds GAL1 and GAL2 with reported Kis of 11.7 μM and 34.1 μM and has demonstrated anticonvulsant and anxiolytic effects. The compound is supplied as a white to off-white solid with high purity and is soluble in DMSO for in vitro use.

  • Selective galanin receptor agonist (GAL1 and GAL2).
  • Demonstrated anticonvulsant and anxiolytic activity in preclinical models.
  • High purity (98.9%) and white to off-white solid appearance.
  • Soluble in DMSO (125 mg/mL); use fresh DMSO and ultrasonic assistance if needed.
  • Recommended storage sealed, protected from moisture and light; frozen solution stability varies by temperature.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.