Learn More
Medchemexpress LLC Galnon (TFA) | 1217448-19-5 | MFCD06798339 | 98.9% | 792.84 g·mol⁻¹ | C42H47F3N4O8 | 25 MG
SDP
Catalog No. 5000248244
Encompass_Preferred
Galnon TFA is a selective, non-peptide agonist of the galanin receptors GAL1 and GAL2, with reported Ki values of 11.7 μM (GAL1) and 34.1 μM (GAL2). It is used in in vitro and in vivo research to study galanin receptor pharmacology and has demonstrated anticonvulsant and anxiolytic effects.
  • Selective galanin receptor agonist with reported Ki values (GAL1 = 11.7 μM, GAL2 = 34.1 μM).
  • Non-peptide small-molecule ligand suitable for receptor pharmacology studies.
  • Demonstrated anticonvulsant and anxiolytic activity in preclinical studies.
  • Supplied at high purity (98.88%) for research use.
  • Available in small research-scale quantities, including 25 mg.
Catalog No. 50-002-48244 Supplier Medchemexpress LLC Supplier No. HY103536A25MG
Galnon TFA is a selective, non-peptide agonist of the galanin receptors GAL1 and GAL2, with reported Ki values of 11.7 μM (GAL1) and 34.1 μM (GAL2). It is used in in vitro and in vivo research to study galanin receptor pharmacology and has demonstrated anticonvulsant and anxiolytic effects.
  • Selective galanin receptor agonist with reported Ki values (GAL1 = 11.7 μM, GAL2 = 34.1 μM).
  • Non-peptide small-molecule ligand suitable for receptor pharmacology studies.
  • Demonstrated anticonvulsant and anxiolytic activity in preclinical studies.
  • Supplied at high purity (98.88%) for research use.
  • Available in small research-scale quantities, including 25 mg.