Learn More
Aviva Systems Biology Corp GALNT14 Antibody - middle region (ARP49834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GALNT14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GALNT14. Alternate names: FLJ12691, FLJ13977, GALNT15, GalNac-T10, GalNac-T14ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60197