Learn More
Aviva Systems Biology Corp Galnt3 Antibody - N-terminal region (ARP46462_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46462_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Galnt3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: MGC124508, Galnt3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-260